Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Diacetoxybenzaldehyde 98.0+%, TCI America™
SDP

Catalog No. p-7127081 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 67727-64-4
Molecular Formula C11H10O5
Molecular Weight (g/mol) 222.20
MDL Number MFCD00143057
InChI Key WYHMNJKAVNPOOR-UHFFFAOYSA-N
PubChem CID 1517488
IUPAC Name 2-(acetyloxy)-5-formylphenyl acetate
SMILES CC(=O)OC1=C(OC(C)=O)C=C(C=O)C=C1

Specifications

Melting Point 53°C
Color White-Yellow
Quantity 5 g
Formula Weight 222.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Diacetoxybenzaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.