Learn More
AdipoGen 3 4-Dihydroxy-L-phenylalanine
SDP
Supplier:  AdipoGen CDXD0465G100
Encompass_Preferred
Chemical. CAS 59-92-7. Formula C9H11NO4. MW 197.19. Synthetic. Metabolic precursor of the neurotransmitters dopamine, norepinephrine noradrenaline, and epinephrine adrenaline, which are collectively known as catecholamines. This amino acid is produced from L-tyrosine by tyrosine hydroxylase and metabolized by catechol-O-methyl transferase COMT. L-DOPA is capable of crossing the blood brain barrier where it is converted to dopamine. Formulations containing L-DOPA have been used to increase dopamine concentrations in the brain as a treatment for Parkinson’s disease and stroke recovery. Mediates neurotrophic factor release by the brain and CNS. Used for the treatment of Parkinson’s disease and dopamine-responsive dystonia. In addition it is used, as a cell adhesion molecule in serum-free cultures of anchorage-dependent mammalian cells, to prevent surfaces from fouling by bonding antifouling polymers to a susceptible substrate or to stain melanocytes.
Catalog No. 50-256-1071
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.