Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3′,4′-Dimethoxyflavone 98.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
200 mg
1 g
Specifications
Specifications
Melting Point | 153°C |
Color | White-Yellow |
Quantity | 200 mg |
Formula Weight | 282.30 |
Percent Purity | ≥98.0% (GC) |
Physical Form | Crystalline Powder |
Chemical Name or Material | 3′,4′-Dimethoxyflavone |
Safety and Handling
RTECSNumber | DJ3012950 |
TSCA | No |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction