Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(4-Hydroxy-3-methoxyphenyl)-1-propanol 98.0+%, TCI America™
SDP

Catalog No. H10111G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 2305-13-7
Molecular Formula C10H14O3
Molecular Weight (g/mol) 182.22
MDL Number MFCD00016571
InChI Key MWOMNLDJNQWJMK-UHFFFAOYSA-N
Synonym Dihydroconiferyl Alcohol
PubChem CID 16822
ChEBI CHEBI:4559
IUPAC Name 4-(3-hydroxypropyl)-2-methoxyphenol
SMILES COC1=CC(CCCO)=CC=C1O

Specifications

Melting Point 61°C
Color White-Yellow
Quantity 1 g
Formula Weight 182.22
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 3-(4-Hydroxy-3-methoxyphenyl)-1-propanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.