Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3,5-Bis(trifluoromethyl)aniline 97.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
10 g
25 g
250 g
Specifications
Specifications
Color | Yellow |
Boiling Point | 85°C |
Quantity | 10 g |
Formula Weight | 229.13 |
Percent Purity | ≥97.0% (GC) |
Physical Form | Liquid |
Chemical Name or Material | 3,5-Bis(trifluoromethyl)aniline |
Safety and Handling
RTECSNumber | ZE9800000 |
TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction