Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Bis(trifluoromethyl)benzoyl Chloride 98.0+%, TCI America™
SDP

Catalog No. p-7119411 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 785-56-8
Molecular Formula C9H3ClF6O
Molecular Weight (g/mol) 276.562
MDL Number MFCD00000387
InChI Key WAKMMQSMEDJRRI-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl benzoyl chloride, 3,5-bis-trifluoromethylbenzoyl chloride, 3,5-di trifluoromethyl benzoyl chloride, 3,5-bis trifluoromethyl benzoylchloride, benzoyl chloride, 3,5-bis trifluoromethyl, 3,5-bis-trifluoromethyl benzoyl chloride, benzoylchloride, 3,5-bis trifluoromethyl, fxffr cvg exfff, 3,5-bis trifluoromethyl benzoychloride
PubChem CID 101856
IUPAC Name 3,5-bis(trifluoromethyl)benzoyl chloride
SMILES C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C(=O)Cl

Specifications

Boiling Point 81°C
Quantity 5 g
UN Number 3265
Formula Weight 276.56
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 3,5-Bis(trifluoromethyl)benzoyl Chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.