Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Diamino-2,4,6-trimethylbenzenesulfonic Acid 93.0+%, TCI America™
SDP

Catalog No. p-7127454 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 32432-55-6
Molecular Formula C9H14N2O3S
Molecular Weight (g/mol) 230.282
MDL Number MFCD00043827
InChI Key PKKGGWLTUCMSSD-UHFFFAOYSA-N
Synonym diaminomesitylenesulfonic acid, benzenesulfonic acid, 3,5-diamino-2,4,6-trimethyl, 2,4-diamino-6-sulfomesitylene, 3,5-diamino-2,4,6-trimethylbenzenesulphonic acid, 2,4,6-trimethyl-3,5-diaminobenzenesulfonic acid, 3,5-diamino-2,4,6-trimethylbenzene sulfonic acid, acmc-209ht1, 2-14-00-00453 beilstein handbook reference, 2,6-diaminomesitylene-4-sulfonic acid, 3,5-diamino-2,4,6-trimethylbenzenesulfonicacid
PubChem CID 94430
IUPAC Name 3,5-diamino-2,4,6-trimethylbenzenesulfonic acid
SMILES CC1=C(C(=C(C(=C1N)C)S(=O)(=O)O)C)N

Specifications

Quantity 5 g
Formula Weight 230.28
Percent Purity ≥93.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Diamino-2,4,6-trimethylbenzenesulfonic Acid
EINECSNumber (3)-2063
RTECSNumber DB5987000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.