Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethyl-1,2,4-triazole 95.0+%, TCI America™
SDP

Catalog No. p-7128296 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 7343-34-2
Molecular Formula C4H7N3
Molecular Weight (g/mol) 97.121
MDL Number MFCD00656686
InChI Key XYYXDARQOHWBPO-UHFFFAOYSA-N
PubChem CID 139001
IUPAC Name 3,5-dimethyl-1H-1,2,4-triazole
SMILES CC1=NC(=NN1)C

Specifications

Melting Point 140°C
Color White-Yellow
Boiling Point 260°C
Quantity 1 g
Formula Weight 97.12
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dimethyl-1,2,4-triazole
EINECSNumber (5)-0600
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.