Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-{[(9H-Fluoren-9-ylmethoxy)carbonyl]amino}-5-hydroxybenzoic acid, ≥97%, Thermo Scientific™

Catalog No. MO07963DA
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 176442-21-0
Molecular Formula C22H17NO5
Molecular Weight (g/mol) 375.38
MDL Number MFCD02682203
InChI Key WGBZDECNYLZYRE-UHFFFAOYSA-N
Synonym 3-9h-fluoren-9-ylmethoxy carbonyl amino-5-hydroxybenzoic acid, 3-9h-fluoren-9-yl methoxy carbonyl amino-5-hydroxybenzoic acid, 3-amino-5-hydroxybenzoic acid,n-fmoc protected, 3-hydroxy-5-9h-fluorene-9-yl methoxy carbonyl amino benzoic acid, benzoic acid,3-9h-fluoren-9-ylmethoxy carbonyl amino-5-hydroxy
PubChem CID 11057866
IUPAC Name 3-(9H-fluoren-9-ylmethoxycarbonylamino)-5-hydroxybenzoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC4=CC(=CC(=C4)C(=O)O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97+%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 375.38
Percent Purity 97%
Chemical Name or Material 3-{[(9H-Fluoren-9-ylmethoxy)carbonyl]amino}-5-hydroxybenzoic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.