Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-2,2,5,5-tetramethyl-1-pyrrolidinyloxy, 99%, Thermo Scientific Chemicals

Catalog No. p-58899
Click to view available options
Quantity:
500 mg
2 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 34272-83-8
Molecular Formula C8H17N2O
Molecular Weight (g/mol) 157.24
MDL Number MFCD00046088
InChI Key USGBLEMQARVRDC-UHFFFAOYSA-N
Synonym 3s-1-hydroxy-2,2,5,5-tetramethylpyrrolidin-3-aminium
PubChem CID 558590
IUPAC Name 1-hydroxy-2,2,5,5-tetramethylpyrrolidin-3-amine
SMILES CC1(CC(C(N1O)(C)C)N)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 45.0°C
Color Orange
Boiling Point 75.0°C to 80.0°C (1.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 500 mg
Formula Weight 157.24
Percent Purity 99%
Physical Form Hygroscopic Crystals
Chemical Name or Material 3-Amino-2,2,5,5-tetramethyl-1-pyrrolidinyloxy, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breath
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.