Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-4-bromo-5-methylpyrazole 98.0+%, TCI America™
SDP

Catalog No. p-7118172 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 1780-72-9
Molecular Formula C4H6BrN3
Molecular Weight (g/mol) 176.017
MDL Number MFCD03412169
InChI Key WZMBDMWFVPKYCF-UHFFFAOYSA-N
PubChem CID 1241299
IUPAC Name 4-bromo-5-methyl-1H-pyrazol-3-amine
SMILES CC1=C(C(=NN1)N)Br

Specifications

Melting Point 120°C
Quantity 1 g
Formula Weight 176.02
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-4-bromo-5-methylpyrazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.