Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-4-iodo-1H-pyrazole, 97%, Thermo Scientific Chemicals

Catalog No. p-4380041
Click to view available options
Quantity:
1 g
5 g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 81542-51-0
Molecular Formula C3H4IN3
Molecular Weight (g/mol) 208.99
InChI Key CPVCVVMJPIOKMN-UHFFFAOYSA-N
Synonym 4-iodo-1h-pyrazol-3-amine, 3-amino-4-iodo-1h-pyrazole, 4-iodo-2h-pyrazol-3-amine, 4-iodo-3-aminopyrazole, 3-amino-4-iodo-pyrazole, 1h-pyrazol-5-amine, 4-iodo
PubChem CID 12831626
IUPAC Name 4-iodo-1H-pyrazol-5-amine
SMILES C1=NNC(=C1I)N

Specifications

Melting Point 143°C to 146°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 208.99
Percent Purity 97%
Chemical Name or Material 3-Amino-4-iodo-1H-pyrazole
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.