Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-4-methylbenzamide 98.0+%, TCI America™
SDP

Catalog No. p-7117865 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 19406-86-1
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.181
MDL Number MFCD00035936
InChI Key VYBKAZXQKUFAHG-UHFFFAOYSA-N
Synonym benzamide, 3-amino-4-methyl, 3-amino-p-toluamide, 3-amino-4-methyl-benzamide, unii-51vb4677ff, atad, acmc-1c8qe, dsstox_cid_24407, dsstox_rid_80206, 3-amino-4-methyl benzamide, 3-azanyl-4-methyl-benzamide
PubChem CID 88043
IUPAC Name 3-amino-4-methylbenzamide
SMILES CC1=C(C=C(C=C1)C(=O)N)N

Specifications

Melting Point 131°C
Quantity 5 g
Formula Weight 150.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-4-methylbenzamide
EINECSNumber (3)-1476
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.