Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-Amino-4-methylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
1 g
5 g
Specifications
Specifications
Chemical Name or Material | 3-Amino-4-methylphenylboronic Acid (contains varying amounts of Anhydride) |
CAS | 22237-12-3 |
Molecular Formula | C7H11BClNO2 |
MDL Number | MFCD01632201 |
Quantity | 1 g |
Synonym | 3-amino-4-methylphenyl boronic acid, 3-amino-4-methylbenzeneboronic acid, 3-amino-4-methyl-phenyl boronic acid, 3-amino-p-tolylboronic acid, boronic acid, 3-amino-4-methylphenyl, acmc-20agxu, pubchem1745, 3-amino-4-methylphenylboranic acid, 3-amino4methyl phenyl boronic acid |
InChI Key | BKFDOPYYSBQWIK-UHFFFAOYSA-N |
SMILES | Cl.CC1=CC=C(C=C1N)B(O)O |
IUPAC Name | (3-amino-4-methylphenyl)boronic acid hydrochloride |
Molecular Weight (g/mol) | 187.43 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction