Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-5-nitrobenz[d]isothiazole 93.0+%, TCI America™
SDP

Catalog No. A15625G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g

Chemical Identifiers

CAS 84387-89-3
Molecular Formula C7H5N3O2S
Molecular Weight (g/mol) 195.196
MDL Number MFCD00270828
InChI Key LDTCWISGJYTXDC-UHFFFAOYSA-N
Synonym 3-amino-5-nitrobenzoisothiazole, 5-nitrobenzo d isothiazol-3-amine, 3-amino-5-nitrobenz d isothiazole, 3-amino-5-nitrobenzisothiazole, 1,2-benzisothiazol-3-amine, 5-nitro, 5-nitro-1,2-benzisothiazol-3-amine, 3-amino-5-nitro-1,2-benzisothiazole, 1,2-benzisothiazol-3-amine,5-nitro, 3-amino-5-nitrobenzo 2,1 isothiazole, 5-nitrobenzo d isothiazole-3-ylamine
PubChem CID 158638
IUPAC Name 5-nitro-1,2-benzothiazol-3-amine
SMILES C1=CC2=C(C=C1[N+](=O)[O-])C(=NS2)N

Specifications

Quantity 5 g
Formula Weight 195.20
Percent Purity ≥93.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-5-nitrobenz[d]isothiazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.