Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-2,5-dichloropyridine 98.0+%, TCI America™
SDP

Catalog No. B48325G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:

Chemical Identifiers

CAS

138006-41-4

Molecular Formula

C5H2BrCl2N

Molecular Weight (g/mol)

226.88

MDL Number

MFCD06659484

InChI Key

CDBHWRZEXFQNBI-UHFFFAOYSA-N

Synonym

2,5-dichloro-3-bromo-pyridine, 2-chloro-3-bromo-5-chloropyridine, 3-bromo-2,5-dichloro-pyridine, pyridine, 3-bromo-2,5-dichloro, 2,5-dichloro-3-bromopyridine, 2,5 dichloro-3-bromo-pyridine, pyridine,3-bromo-2,5-dichloro, pubchem1173, acmc-209cf8, 3-bromo-2, 5-dichloropyridine

PubChem CID

12901596

IUPAC Name

3-bromo-2,5-dichloropyridine

SMILES

ClC1=CC(Br)=C(Cl)N=C1

Specifications

Melting Point

39°C

Color

Yellow

Quantity

5 g

Formula Weight

226.88

Percent Purity

≥98.0% (GC)

Physical Form

Crystalline Powder

Chemical Name or Material

3-Bromo-2,5-dichloropyridine

TSCA

No

Recommended Storage

Refrigerator

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.