Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-2,5-dichloropyridine 98.0+%, TCI America™
SDP

Catalog No. p-7123779 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 138006-41-4
Molecular Formula C5H2BrCl2N
Molecular Weight (g/mol) 226.88
MDL Number MFCD06659484
InChI Key CDBHWRZEXFQNBI-UHFFFAOYSA-N
Synonym 2,5-dichloro-3-bromo-pyridine, 2-chloro-3-bromo-5-chloropyridine, 3-bromo-2,5-dichloro-pyridine, pyridine, 3-bromo-2,5-dichloro, 2,5-dichloro-3-bromopyridine, 2,5 dichloro-3-bromo-pyridine, pyridine,3-bromo-2,5-dichloro, pubchem1173, acmc-209cf8, 3-bromo-2, 5-dichloropyridine
PubChem CID 12901596
IUPAC Name 3-bromo-2,5-dichloropyridine
SMILES ClC1=CC(Br)=C(Cl)N=C1

Specifications

Melting Point 39°C
Color Yellow
Quantity 1 g
Formula Weight 226.88
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-2,5-dichloropyridine
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.