Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-4-fluorotoluene 98.0+%, TCI America™
SDP

Catalog No. B19455G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g

Chemical Identifiers

CAS 452-62-0
Molecular Formula C7H6BrF
Molecular Weight (g/mol) 189.03
MDL Number MFCD00040827
InChI Key QLRKALMVPCQTMU-UHFFFAOYSA-N
Synonym 3-bromo-4-fluorotoluene, benzene, 2-bromo-1-fluoro-4-methyl, 3-bromo-4-fluoro toluene, 2-bromo-1-fluoro-4-methyl-benzene, 2-brom-1-fluor-4-methylbenzol, pubchem1600, 4-fluoro-3-bromotoluene, acmc-1aei5, 3-bromo-4-fluoro-toluene, ksc237s0d
PubChem CID 96745
IUPAC Name 2-bromo-1-fluoro-4-methylbenzene
SMILES CC1=CC=C(F)C(Br)=C1

Specifications

Color Yellow
Boiling Point 186°C
Quantity 5 g
Formula Weight 189.03
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromo-4-fluorotoluene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.