Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-4-hydroxy-5-methoxybenzonitrile 98.0+%, TCI America™
SDP

Catalog No. p-7123728 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 52805-45-5
Molecular Formula C8H6BrNO2
Molecular Weight (g/mol) 228.045
MDL Number MFCD01123027
InChI Key FHXLXRNDUJYBEQ-UHFFFAOYSA-N
Synonym 3-bromo-4-hydroxy-5-methoxy-benzonitrile, 3-bromo-4-hydroxy-5-methoxybenzenecarbonitrile, acmc-1atrh, 3-bromo-4-hydroxy-5-methoxybenzonitrile
PubChem CID 7021625
IUPAC Name 3-bromo-4-hydroxy-5-methoxybenzonitrile
SMILES COC1=C(C(=CC(=C1)C#N)Br)O

Specifications

Melting Point 146°C
Color White
Quantity 1 g
UN Number 3439
Formula Weight 228.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-4-hydroxy-5-methoxybenzonitrile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.