Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-4-methoxyphenylacetic Acid 98.0+%, TCI America™
SDP

Catalog No. p-7123686 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 774-81-2
Molecular Formula C9H9BrO3
Molecular Weight (g/mol) 245.072
MDL Number MFCD00017538
InChI Key POTVGQUUEQTPNA-UHFFFAOYSA-N
PubChem CID 136615
IUPAC Name 2-(3-bromo-4-methoxyphenyl)acetic acid
SMILES COC1=C(C=C(C=C1)CC(=O)O)Br

Specifications

Melting Point 117°C
Color White-Yellow
Quantity 1 g
UN Number 3077
Formula Weight 245.07
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-4-methoxyphenylacetic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.