Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-5-isocyanatopyridine, 95%, Tech., Thermo Scientific™

Catalog No. p-3586717
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 167951-51-1
Molecular Formula C6H3BrN2O
Molecular Weight (g/mol) 199.007
MDL Number MFCD09879965
InChI Key PNEYUZGELZQGCX-UHFFFAOYSA-N
PubChem CID 24229746
IUPAC Name 3-bromo-5-isocyanatopyridine
SMILES C1=C(C=NC=C1Br)N=C=O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 5-bromo-3-pyridyl isocyanate, pyridine,3-bromo-5-isocyanato, pyridine, 3-bromo-5-isocyanato, 5-bromopyridin-3-isocyanate, 3-bromo-5-isocyanatopyridine,tech
Formula Weight 199.01
Percent Purity 95%
Grade Technical
Chemical Name or Material 3-Bromo-5-isocyanatopyridine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.