Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-5-nitroanisole 98.0+%, TCI America™
SDP

Catalog No. p-7123876 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 16618-67-0
Molecular Formula C7H6BrNO3
Molecular Weight (g/mol) 232.033
MDL Number MFCD04112752
InChI Key MEQKSFQEPDRNEQ-UHFFFAOYSA-N
Synonym 3-bromo-5-nitroanisole, 3-bromo-5-nitroanisol, 5-bromo-1-methoxy-3-nitrobenzene, 1-bromo-3-methoxy-5-nitro-benzene, benzene, 1-bromo-3-methoxy-5-nitro, pubchem21624, acmc-209dts, 3-bromo-5-methoxynitrobenzene, benzene,1-bromo-3-methoxy-5-nitro, 3-methoxy-5-nitrophenyl bromide
PubChem CID 12299076
IUPAC Name 1-bromo-3-methoxy-5-nitrobenzene
SMILES COC1=CC(=CC(=C1)[N+](=O)[O-])Br

Specifications

Melting Point 88°C
Color Yellow
Quantity 1 g
Formula Weight 232.03
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-5-nitroanisole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.