Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-5-nitrobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. p-7122905 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 6307-83-1
Molecular Formula C7H4BrNO4
Molecular Weight (g/mol) 246.016
MDL Number MFCD00100098
InChI Key AXRKIZCFYZBBPX-UHFFFAOYSA-N
Synonym 3-bromo-5-nitrobenzoic acid, 5-bromo-3-nitrobenzoic acid, benzoic acid, 3-bromo-5-nitro, 3-bromo-5-nitro-benzoic acid, 3-bromo-5-nitrobenzoicacid, pubchem4734, acmc-209ncw, ksc357s4t, 3-bromo-5-nitro benzoic acid
PubChem CID 239336
SMILES C1=C(C=C(C=C1[N+](=O)[O-])Br)C(=O)O

Specifications

Melting Point 162°C
Quantity 1 g
Formula Weight 246.02
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-5-nitrobenzoic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.