Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-5-(trifluoromethyl)benzoic Acid 98.0+%, TCI America™
SDP

Catalog No. p-7123599 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 328-67-6
Molecular Formula C8H4BrF3O2
Molecular Weight (g/mol) 269.02
MDL Number MFCD03412186
InChI Key AMZBKZQMAZWIJM-UHFFFAOYSA-N
Synonym 3-bromo-5-trifluoromethyl benzoic acid, 3-bromo-5-trifluoromethyl-benzoic acid, 5-bromo-3-trifluoromethyl benzoic acid, 3-bromo-5-trifluoromethylbenzoicacid, benzoic acid, 3-bromo-5-trifluoromethyl, pubchem4737, 3-bromo-5-trifluoromethyl benzoicacid, acmc-1clgk, ksc222e0t
PubChem CID 11086788
IUPAC Name 3-bromo-5-(trifluoromethyl)benzoic acid
SMILES OC(=O)C1=CC(=CC(Br)=C1)C(F)(F)F

Specifications

Melting Point 135°C
Color White-Yellow
Quantity 5 g
Formula Weight 269.02
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-5-(trifluoromethyl)benzoic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.