Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromophenyl Methyl Sulfone 98.0+%, TCI America™
SDP

Catalog No. p-7123229 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 34896-80-5
Molecular Formula C7H7BrO2S
Molecular Weight (g/mol) 235.10
MDL Number MFCD03095326
InChI Key WBOMXUMQOVQNKT-UHFFFAOYSA-N
PubChem CID 2757146
IUPAC Name 1-bromo-3-methanesulfonylbenzene
SMILES CS(=O)(=O)C1=CC=CC(Br)=C1

Specifications

Melting Point 105°C
Color White-Yellow
Quantity 5 g
Formula Weight 235.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromophenyl Methyl Sulfone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.