Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Butoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. p-7193857 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 352534-81-7
Molecular Formula C10H15BO3
Molecular Weight (g/mol) 194.037
MDL Number MFCD03427053
InChI Key ZTRKYYYDXARQRD-UHFFFAOYSA-N
Synonym 3-Butoxybenzeneboronic Acid
PubChem CID 5084101
IUPAC Name (3-butoxyphenyl)boronic acid
SMILES B(C1=CC(=CC=C1)OCCCC)(O)O

Specifications

Melting Point 93°C
Color White
Quantity 1 g
Formula Weight 194.04
Physical Form Crystalline Powder
Chemical Name or Material 3-Butoxyphenylboronic Acid (contains varying amounts of Anhydride)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.