Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-2,2,3,3-tetrafluoropropionic Acid 98.0+%, TCI America™
SDP

Catalog No. C23565G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g

Chemical Identifiers

CAS 661-82-5
Molecular Formula C3HClF4O2
Molecular Weight (g/mol) 180.483
MDL Number MFCD00153099
InChI Key XJRMCFMZGRJXEZ-UHFFFAOYSA-N
PubChem CID 2782491
IUPAC Name 3-chloro-2,2,3,3-tetrafluoropropanoic acid
SMILES C(=O)(C(C(F)(F)Cl)(F)F)O

Specifications

Color Yellow
Boiling Point 131°C
Quantity 5 g
UN Number 3265
Formula Weight 180.48
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 3-Chloro-2,2,3,3-tetrafluoropropionic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.