Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-chloro-2-methylphenyl isothiocyanate, 97%, Thermo Scientific™

Catalog No. TL00697DA
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 19241-35-1
Molecular Formula C8H6ClNS
Molecular Weight (g/mol) 183.653
MDL Number MFCD00022056
InChI Key ZXEZATIRZLJXFU-UHFFFAOYSA-N
PubChem CID 140504
IUPAC Name 1-chloro-3-isothiocyanato-2-methylbenzene
SMILES CC1=C(C=CC=C1Cl)N=C=S

Specifications

CAS Min % 97.0
Quantity 1 g
Packaging Amber Glass Bottle
Assay Percent Range 97%
Synonym 3-chloro-2-methylphenyl isothiocyanate, isothiocyanic acid, 3-chloro-o-tolyl ester, 3-chloro-2-methylphenylisothiocyanate, benzene,1-chloro-3-isothiocyanato-2-methyl, 3-chloro-2-methylbenzenisothiocyanate, pubchem10477, 3-chloro-2-methylphenyl-isothiocyanate, 1-chloranyl-3-isothiocyanato-2-methyl-benzene, benzene, 1-chloro-3-isothiocyanato-2-methyl
Formula Weight 183.66
Percent Purity 97%
Chemical Name or Material 3-Chloro-2-methylphenyl isothiocyanate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.
Causes serious eye irritation.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.