Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-4-hydroxybenzoic Acid Hemihydrate 98.0+%, TCI America™
SDP

Catalog No. p-7124808 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
250 g

Chemical Identifiers

CAS 3964-58-7
Molecular Formula C7H5ClO3
Molecular Weight (g/mol) 172.56
MDL Number MFCD00002549
InChI Key QGNLHMKIGMZKJX-UHFFFAOYSA-N
Synonym benzoic acid, 3-chloro-4-hydroxy, 3-chloro-4-hydroxy-benzoic acid, hemihydrate, pubchem13038, acmc-209vmh, 3-chloro-4hydroxybenzoic acid, 3-chloro4-hydroxybenzoic acid, 4-hydroxy-3-chlorobenzoic acid, 3-chloro-4-hydroxybenzoic scid, 3-chloro-4-hydroxy benzoic acid
PubChem CID 19860
IUPAC Name 3-chloro-4-hydroxybenzoic acid
SMILES OC(=O)C1=CC=C(O)C(Cl)=C1

Specifications

Melting Point 171°C
Color White
Quantity 25 g
Formula Weight 172.56
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-4-hydroxybenzoic Acid Hemihydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.