Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-4-methoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. p-7193877 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 175883-60-0
Molecular Formula C7H8BClO3
Molecular Weight (g/mol) 186.40
MDL Number MFCD04039888
InChI Key VUTJHOWRPUPHIG-UHFFFAOYSA-N
Synonym 3-chloro-4-methoxyphenyl boronic acid, 3-chloro-4-methoxybenzeneboronic acid, 4-borono-2-chloroanisole, 3-chloro-4-methoxyphenyl boranediol, boronic acid, 3-chloro-4-methoxyphenyl, akos brn-0653, 3-chloro-4, acmc-1c1io, ksc489k9h, 3-chloro-4-methoxyphenyl boronicacid
PubChem CID 3247522
IUPAC Name (3-chloro-4-methoxyphenyl)boronic acid
SMILES COC1=C(Cl)C=C(C=C1)B(O)O

Specifications

Melting Point 242°C
Color White
Quantity 1 g
Formula Weight 186.40
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-4-methoxyphenylboronic Acid (contains varying amounts of Anhydride)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.