Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-5-(trifluoromethyl)pyridine 97.0+%, TCI America™
SDP

Catalog No. p-7125741 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 85148-26-1
Molecular Formula C6H3ClF3N
Molecular Weight (g/mol) 181.54
MDL Number MFCD00042227
InChI Key OMRCXTBFBBWTDL-UHFFFAOYSA-N
Synonym 3-chloro-5-trifluoromethyl pyridine, 3-trifluoromethyl-5-chloropyridine, pyridine, 3-chloro-5-trifluoromethyl, pubchem3552, acmc-209zee, ksc495c1f, 3-chloro-5-trifluoromethyl-pyridine, 3-chloro-5-trifluoromethyl pyridine-carbonitrile
PubChem CID 2736698
IUPAC Name 3-chloro-5-(trifluoromethyl)pyridine
SMILES FC(F)(F)C1=CC(Cl)=CN=C1

Specifications

Color Yellow
Boiling Point 138°C
Quantity 1 g
UN Number 1992
Formula Weight 181.54
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chloro-5-(trifluoromethyl)pyridine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.