Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloropyrazine-2-carbonitrile, 95+%, Thermo Scientific™

Catalog No. KM04911DA
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 55557-52-3
Molecular Formula C5H2ClN3
Molecular Weight (g/mol) 139.54
MDL Number MFCD00219653
InChI Key SDLFAEGTVBPHBK-UHFFFAOYSA-N
Synonym 2-chloro-3-cyanopyrazine, 3-chloro-2-pyrazinecarbonitrile, 2-chloro-3-cyannopyrazine, pyrazinecarbonitrile, 3-chloro, 3-chloro-2-cyanopyrazine, 3-chlorpyrazin-2-carbonitril, chlorocyanopyrazine, pubchem10180, acmc-209lnr, 2-cyano-3-chloropyrazine
PubChem CID 292465
IUPAC Name 3-chloropyrazine-2-carbonitrile
SMILES ClC1=NC=CN=C1C#N

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 139.54
Percent Purity 97%
Chemical Name or Material 3-Chloropyrazine-2-carbonitrile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.