Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chlorothiophene 97.0+%, TCI America™
SDP

Catalog No. p-7124847 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 17249-80-8
Molecular Formula C4H3ClS
Molecular Weight (g/mol) 118.578
MDL Number MFCD00043887
InChI Key QUBJDMPBDURTJT-UHFFFAOYSA-N
Synonym thiophene, 3-chloro, 3-chloro thiophene, 3-thienyl chloride, 3-chlorthiophen, 3-chloro-thiophene, 3-chloranylthiophene, pubchem5477, acmc-1cdmv, 3-chlorothiophene, ksc175g2h
PubChem CID 87017
IUPAC Name 3-chlorothiophene
SMILES C1=CSC=C1Cl

Specifications

Color Yellow
Boiling Point 139°C
Quantity 5 g
UN Number 1993
Formula Weight 118.58
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chlorothiophene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.