Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3'-Deoxy-2',3'-didehydrothymidine, 98%, Thermo Scientific Chemicals

Catalog No. p-7077523
Click to view available options
Quantity:
1 g
5 g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3056-17-5
Molecular Formula C10H12N2O4
Molecular Weight (g/mol) 224.22
MDL Number MFCD00132921
InChI Key XNKLLVCARDGLGL-HGXVMFPFNA-N
Synonym stavudine, sanilvudine, zerit, 2',3'-didehydro-3'-deoxythymidine, estavudina, stavudinum, zerit xr, zerut xr, stavudinum inn-latin, 2',3'-anhydrothymidine
PubChem CID 18283
ChEBI CHEBI:63581
IUPAC Name 1-[(2R,5S)-5-(hydroxymethyl)-2,5-dihydrofuran-2-yl]-5-methylpyrimidine-2,4-dione
SMILES CC1=CN([C@@H]2O[C@H](CO)C=C2)C(=O)NC1=O

Specifications

Melting Point 162.0°C to 169.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Merck Index 158929
Formula Weight 224.21
Percent Purity 98%
Chemical Name or Material 3'-Deoxy-2',3'-didehydrothymidine
Hazard Category Serious eye damage/eye irritation (category 2), Germ cell mutagenicity (category 2), Specific target organ toxicity after single exposure (category 3), Skin corrosion/irritation (category 2)
Hazard Statement Causes serious eye irritation, Suspected of causing genetic defects, May cause respiratory irritation, Causes skin irritation

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.