Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-[[Dimethyl(vinyl)silyl]oxy]-1,1,5,5-tetramethyl-3-phenyl-1,5-divinyltrisiloxane 98.0+%, TCI America™
SDP

Catalog No. p-7128715 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 60111-47-9
Molecular Formula C18H32O3Si4
Molecular Weight (g/mol) 408.791
MDL Number MFCD00156495
InChI Key XYVYGTWMOAIWOG-UHFFFAOYSA-N
Synonym Tris[dimethyl(vinyl)siloxy]phenylsilane
PubChem CID 6453907
IUPAC Name tris[[ethenyl(dimethyl)silyl]oxy]-phenylsilane
SMILES C[Si](C)(C=C)O[Si](C1=CC=CC=C1)(O[Si](C)(C)C=C)O[Si](C)(C)C=C

Specifications

Color Colorless
Quantity 5 g
Formula Weight 408.79
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-[[Dimethyl(vinyl)silyl]oxy]-1,1,5,5-tetramethyl-3-phenyl-1,5-divinyltrisiloxane
EINECSNumber (7)-0481
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.