Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Ethyl-2-methylbenzoxazolium Iodide 98.0+%, TCI America™
SDP

Catalog No. p-7129022 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 5260-37-7
Molecular Formula C10H12INO
Molecular Weight (g/mol) 289.12
MDL Number MFCD00044141
InChI Key UZWYLWSUBQEUMC-UHFFFAOYSA-M
Synonym 3-ethyl-2-methylbenzoxazolium iodide, 3-ethyl-2-methylbenzo d oxazol-3-ium iodide, 2-methyl-3-ethylbenzoxazolium iodide, benzoxazolium, 3-ethyl-2-methyl-, iodide, benzoxazolium, 3-ethyl-2-methyl-, iodide 1:1, 3-ethyl-2-methyl-1,3-benzoxazol-3-ium iodide, 3-ethyl-2-methylbenzoxazoliumiodide, acmc-209l0h, ethyl-2-methyl-benzoxazolium iodide, 3-ethyl-2-methylbenzoxazolium iodide, technical grade
PubChem CID 21331
IUPAC Name 3-ethyl-2-methyl-1,3-benzoxazol-3-ium iodide
SMILES [I-].CC[N+]1=C(C)OC2=CC=CC=C12

Specifications

Melting Point 197°C
Quantity 5 g
Formula Weight 289.12
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Ethyl-2-methylbenzoxazolium Iodide
EINECSNumber (5)-0159
RTECSNumber DM5285000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.