Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Ethylamino)phenol Hemisulfate 98.0+%, TCI America™
SDP

Catalog No. p-7129052 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 1274892-48-6
Molecular Formula C16H24N2O6S
Molecular Weight (g/mol) 372.436
MDL Number MFCD03093630
InChI Key CLHKUUQJWJTXSF-UHFFFAOYSA-N
PubChem CID 44630284
IUPAC Name 3-(ethylamino)phenol;sulfuric acid
SMILES CCNC1=CC(=CC=C1)O.CCNC1=CC(=CC=C1)O.OS(=O)(=O)O

Specifications

Color Gray-White
Quantity 1 g
Formula Weight 186.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Ethylamino)phenol Hemisulfate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.