Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3'-Formyl(1,1'-biphenyl)-3-carboxylic acid, 97%, Thermo Scientific™

Catalog No. MO01257CB
Click to view available options
Quantity:
250 mg

Chemical Identifiers

CAS 222180-19-0
Molecular Formula C14H10O3
Molecular Weight (g/mol) 226.231
MDL Number MFCD04039068
InChI Key QOCUSJPHQSKJJR-UHFFFAOYSA-N
Synonym 3'-formyl-1,1'-biphenyl-3-carboxylic acid, 3-3-formylphenyl benzoic acid, 3'-formylbiphenyl-3-carboxylic acid, 3'-formyl 1,1'-biphenyl-3-carboxylic acid, 3'-formyl-biphenyl-3-carboxylic acid, amtda124, acmc-1cb16, 1,1'-biphenyl-3-carboxylicacid, 3'-formyl
PubChem CID 2794721
IUPAC Name 3-(3-formylphenyl)benzoic acid
SMILES C1=CC(=CC(=C1)C=O)C2=CC(=CC=C2)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 226.23
Percent Purity 97%
Chemical Name or Material 3'-Formyl(1, 1'-biphenyl)-3-carboxylic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.