Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-Glycidyloxypropyl(dimethoxy)methylsilane 96.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
25 g
500 g
Specifications
Specifications
Color | Yellow |
Boiling Point | 100°C |
Quantity | 25 g |
Formula Weight | 220.34 |
Percent Purity | ≥96.0% (GC) |
Physical Form | Liquid |
Chemical Name or Material | 3-Glycidyloxypropyl(dimethoxy)methylsilane |
Safety and Handling
EINECSNumber | (2)-2072 |
TSCA | No |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction