Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxy-4-iodobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. p-7131074 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 58123-77-6
Molecular Formula C7H5IO3
Molecular Weight (g/mol) 264.02
MDL Number MFCD02068386
InChI Key UABBBWVTEWIIMN-UHFFFAOYSA-N
Synonym 3-hydroxy-4-iodo-benzoic acid, benzoic acid, 3-hydroxy-4-iodo, acmc-1ay71, ksc497o7h, 4-iodo-3-hydroxybenzoic acid, 4-iodo-3-hydroxy-benzoic acid, 3-hydroxy-4-iodobenzoic acid, 4-iodo-3-hydroxy benzoic acid
PubChem CID 736854
IUPAC Name 3-hydroxy-4-iodobenzoic acid
SMILES OC(=O)C1=CC(O)=C(I)C=C1

Specifications

Melting Point 226°C
Color White
Quantity 1 g
Formula Weight 264.02
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-4-iodobenzoic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.