Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxy-alpha-methylbenzyl Alcohol 98.0+%, TCI America™
SDP

Catalog No. p-7131042 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 2415-09-0
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00238621
InChI Key COJRWHSKVYUZHQ-UHFFFAOYSA-N
Synonym 1-(3-Hydroxyphenyl)ethanol, 3-(1-Hydroxyethyl)phenol
PubChem CID 13542886
IUPAC Name 3-(1-hydroxyethyl)phenol
SMILES CC(C1=CC(=CC=C1)O)O

Specifications

Melting Point 118°C
Color White-Yellow
Quantity 1 g
Formula Weight 138.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-alpha-methylbenzyl Alcohol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.