Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Iodo-2,4,5-trimethylthiophene (stabilized with Copper chip) 96.0+%, TCI America™
SDP

Catalog No. p-7131410 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 60813-84-5
Molecular Formula C7H9IS
Molecular Weight (g/mol) 252.113
MDL Number MFCD00191528
InChI Key FYPOBSHIONYIKX-UHFFFAOYSA-N
PubChem CID 12278343
IUPAC Name 3-iodo-2,4,5-trimethylthiophene
SMILES CC1=C(SC(=C1I)C)C

Specifications

Boiling Point 87°C
Quantity 1 g
Formula Weight 252.11
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Iodo-2,4,5-trimethylthiophene (stabilized with Copper chip)
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.