Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Iodoaniline 99.0+%, TCI America™
SDP

Catalog No. p-7131177 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 626-01-7
Molecular Formula C6H6IN
Molecular Weight (g/mol) 219.025
MDL Number MFCD00007781
InChI Key FFCSRWGYGMRBGD-UHFFFAOYSA-N
Synonym m-iodoaniline, benzenamine, 3-iodo, aniline, m-iodo, m-aminoiodobenzene, 3-iodobenzenamine, 1-amino-3-iodobenzene, 3-iodo-phenylamine, 3iodoaniline, 3-iodo-aniline, 3-iodophenylamine
PubChem CID 12271
IUPAC Name 3-iodoaniline
SMILES C1=CC(=CC(=C1)I)N

Specifications

Color White-Yellow
Boiling Point 116°C
Quantity 5 g
Formula Weight 219.03
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 3-Iodoaniline
RTECSNumber BY3750000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.