Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Iodobenzylamine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. p-7131555 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 3718-88-5
Molecular Formula C7H9ClIN
Molecular Weight (g/mol) 269.51
MDL Number MFCD00012857
InChI Key PYFDZOCGFHIRST-UHFFFAOYSA-N
Synonym 3-iodobenzylamine hydrochloride, 3-iodophenyl methanamine hydrochloride, 3-iodobenzylamine hcl, 3-iodobenzylaminehydrochloride, benzenemethanamine, 3-iodo-, hydrochloride, 1-3-iodophenyl methanamine hydrochloride, pubchem3302, acmc-1cmc7, 3-iodobenzylamine hcl salt, ksc491e3h
PubChem CID 2723861
IUPAC Name (3-iodophenyl)methanamine;hydrochloride
SMILES C1=CC(=CC(=C1)I)CN.Cl

Specifications

Melting Point 194°C
Color White-Yellow
Quantity 1 g
Formula Weight 269.51
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Iodobenzylamine Hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.