Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Iodothiophene, 97%, Thermo Scientific Chemicals

Catalog No. AC443380050
Click to view available options
Quantity:

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS

10486-61-0

Molecular Formula

C4H3IS

Molecular Weight (g/mol)

210.03

MDL Number

MFCD00037901

InChI Key

WGKRMQIQXMJVFZ-UHFFFAOYSA-N

Synonym

thiophene, 3-iodo, 3-iodothiophene, stab. with copper, 3-lodothiophene, 3-iodo-thiophene, 3-iodanylthiophene, 3-thienyl iodide, thien-3-yl iodide, pubchem16213, acmc-2098dn

PubChem CID

66332

IUPAC Name

3-iodothiophene

SMILES

IC1=CSC=C1

Specifications

Melting Point

-13.0°C

Density

2.0660g/mL

Boiling Point

195.0°C

Flash Point

78°C

Assay Percent Range

96% min. (GC)

Quantity

5 g

Specific Gravity

2.066

Formula Weight

210.03

Percent Purity

97%

Chemical Name or Material

3-Iodothiophene

Hazard Category

GHS Signal Word: Warning

Hazard Statement

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Precautionary Statement

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unw

EINECSNumber

234-009-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.