Learn More
TARGETMOL CHEMICALS INC 3-METHOXY-DL-TYROSINE 50MG
Supplier:  TARGETMOL CHEMICALS INC T9492
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg and bulk. Please contact Fisher for quotes. 3-METHOXY-DL-TYROSINE is a naturally occurring non-essential amino acid found in the human body and many other organisms. It is a precursor to the neurotransmitter dopamine and is involved in the regulation of dopamine levels in the brain with potential therapeutic applications in a variety of disorders including Parkinson's disease depression and obesity. purity: 99%
Catalog No. 50-257-9004
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.