Learn More
Medchemexpress LLC HY-103638A 100mg Medchemexpress, 3-Methoxytyramine CAS:554-52-9 Purity:>98%
Supplier: Medchemexpress LLC HY103638A100MG
Medchemexpress, HY-103638A 100mg 3-Methoxytyramine CAS:554-52-9 3-Methoxytyramine, a well known extracellular metabolite of 3-hydroxytyramine/dopamine, is a neuromodulator. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.