Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-O-methyl-D-glucose, 99.1%, MP Biomedicals™

Catalog No. p-4404716 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
1 g
10 g

Chemical Identifiers

CAS 13224-94-7
Molecular Formula C7H14O6
Molecular Weight (g/mol) 194.18
MDL Number MFCD00063266
InChI Key SCBBSJMAPKXHAH-OVHBTUCOSA-N
Synonym 3-o-methyl-alpha-d-glucopyranose, 3-o-methyl-a-d-glucopyranose, 2s,3r,4s,5r,6r-6-hydroxymethyl-4-methoxytetrahydro-2h-pyran-2,3,5-triol, 2s,3r,4s,5r,6r-6-hydroxymethyl-4-methoxyoxane-2,3,5-triol, 3-o-methyl-, a-d-glucopyranose, 3-o-methyl-alpha-d-glucose, a-d-glucopyranose,3-o-methyl, alpha-d-glucopyranose, 3-o-methyl, 3-o-methyl-d-glucopyranose
PubChem CID 83246
IUPAC Name (2S,3R,4S,5R,6R)-6-(hydroxymethyl)-4-methoxyoxane-2,3,5-triol
SMILES CO[C@@H]1[C@@H](O)[C@@H](O)O[C@H](CO)[C@H]1O

Specifications

Color White
Quantity 1 g
Formula Weight 194.2
Percent Purity 0.991
Physical Form Powder
Chemical Name or Material 3-O-methyl-D-glucose
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.