Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Phenylisoxazole-5-carboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. p-7135557 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 14442-12-7
Molecular Formula C10H7NO3
Molecular Weight (g/mol) 189.17
MDL Number MFCD01936010
InChI Key YGTFDJRCCBKLDM-UHFFFAOYSA-N
Synonym 3-phenylisoxazole-5-carboxylic acid, 3-phenyl-5-isoxazolecarboxylic acid, 5-isoxazolecarboxylic acid, 3-phenyl, 3-phenyl-isoxazole-5-carboxylic acid, phenylisoxazolecarboxylicacid, 5-carboxy-3-phenylisoxazole, 3-phenylisoxazole-5-carboxylic, 3-phenyl-5-isoxazolecarboxylicacid, 3-phenylisoxazole-5-carboxylicacid, 3-phenyl-5-isoxazole carboxylic acid
PubChem CID 737474
IUPAC Name 3-phenyl-1,2-oxazole-5-carboxylic acid
SMILES C1=CC=C(C=C1)C2=NOC(=C2)C(=O)O

Specifications

Melting Point 178°C
Color Yellow
Quantity 1 g
Formula Weight 189.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Phenylisoxazole-5-carboxylic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.