Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Pyridinecarboxamide Oxime 98.0+%, TCI America™
SDP

Catalog No. p-7135354 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 1594-58-7
Molecular Formula C6H7N3O
Molecular Weight (g/mol) 137.14
MDL Number MFCD00265955
InChI Key AQBMQGDKWIPBRF-UHFFFAOYSA-N
Synonym 3-pyridylamidoxime, 3-pyridinecarboxamidoxime, z-n'-hydroxynicotinimidamide, n-hydroxynicotinimidamide, z-n'-hydroxypyridine-3-carboximidamide, n-hydroxy-nicotinamidine, 3-pyridinecarboxamidoxine, 3-pyridinecarboxamide oxime, nicotinamidoxime, hydroxyimino-3-pyridylmethylamine
PubChem CID 5372334
IUPAC Name (Z)-N'-hydroxypyridine-3-carboximidamide
SMILES N\C(=N/O)C1=CC=CN=C1

Specifications

Melting Point 132°C
Color White-Yellow
Quantity 1 g
Formula Weight 137.14
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Pyridinecarboxamide Oxime
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.